missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PNLIP Protein
A DNA sequence encoding the Mus musculus (Mouse) PNLIP, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_081201.2 |
---|---|
Gene ID (Entrez) | 69060 |
Name | PNLIP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16030651
|
enQuireBio™
QP7762-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16050651
|
enQuireBio™
QP7762-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16040651
|
enQuireBio™
QP7762-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16060651
|
enQuireBio™
QP7762-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16070651
|
enQuireBio™
QP7762-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16080651
|
enQuireBio™
QP7762-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_081201.2 | |
PNLIP Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
69060 | |
Research Use Only | |
Pnlip | |
Recombinant Protein | |
E. coli | |
REVCFDKLGC FSDDAPWSGT LDRPLKALPW SPAQINTRFL LYTNENPDNY QLITSDASNI RNSNFRTNRK TRIIIHGFID KGEENWLSDM CKNMFRVESV NCICVDWKGG SRTTYTQATQ NVRVVGAEVA LLVNVLQSDL GYSLNNVHLI GHSLGSHIAG EAGKRTFGAI GRITGLDPAE PYFQGTPEEV RLDPTDAQFV DAIHTDAGPI IPNLGFGMSQ TVGHLDFFPN GGIEMPGCQK NILSQIVDID GIWEGTRNFA ACNHLRSYKF YTDSIVNPTG FAGFSCSSYS LFTANKCFPC GSGGCPQMGH YADRYPGKTS RLYQTFYLNT GDKSNFARWR YQVTVTLSGQ KVTGHILVSL FGNGGNSKQY EVFKGSLQPG TSHVNEFDSD VDVGDLQKVK FIWYNNVINP TLPKVGASRI TVERNDGRVF NFCSQETVRE DVLLTLSPC | |
Greater than 90% as determined by SDS-PAGE. |