missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Major urinary protein 1
A DNA sequence encoding the Mus musculus (Mouse) Major urinary protein 2, was expressed in the hosts and tags indicated.
245.00€ - 2645.00€
Specifications
Name | Major urinary protein 1 |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | Mup2 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017150
|
enQuireBio™
QP7038-YE-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16057150
|
enQuireBio™
QP7038-YE-500UG |
500 μg |
2195.00€
500µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16027150
|
enQuireBio™
QP7038-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16047150
|
enQuireBio™
QP7038-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16067150
|
enQuireBio™
QP7038-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16037150
|
enQuireBio™
QP7038-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
Major urinary protein 1 | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
Mup2 | |
Recombinant Protein | |
Yeast | |
EEASSTGRNF NVEKINGEWH TIILASDKRE KIEDNGNFRL FLEQIHVLEK SLVLKFHTVR DEECSELSMV ADKTEKAGEY SVTYDGFNTF TIPKTDYDNF LMAHLINEKD GETFQLMGLY GREPDLSSDI KERFAKLCEE HGILRENIID LSNANRCLQA RE | |
Greater than 90% as determined by SDS-PAGE. |