missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Interferon epsilon Protein
A DNA sequence encoding the Mus musculus (Mouse) Interferon epsilon, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7798-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_796322.1 | |
Interferon epsilon Protein | |
Research Use Only | |
Ifne | |
Recombinant Protein | |
Yeast | |
LEPKRIPFQL WMNRESLQLL KPLPSSSVQQ CLAHRKNFLL PQQPVSPHQY QEGQVLAVVH EILQQIFTLL QTHGTMGIWE ENHIEKVLAA LHRQLEYVES LGGLNAAQKS GGSSAQNLRL QIKAYFRRIH DYLENQRYSS CAWIIVQTEI HRCMFFVFRF TTWLSRQDPD P | |
Greater than 90% as determined by SDS-PAGE. |
230405 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |