missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Insulin-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Insulin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9628-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032412.3 | |
Insulin-1 Protein | |
Research Use Only | |
Ins1 | |
Recombinant Protein | |
Yeast | |
FVKQHLCGPH LVEALYLVCG ERGFFYTPKS RREVEDPQVE QLELGGSPGD LQTLALEVAR QKRGIVDQCC TSICSLYQLE NYCN | |
Greater than 80% as determined by SDS-PAGE. |
16333 | |
100 μg | |
Not determined. | |
Full. Length. of. Mature. Protein. | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |