missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL-1 beta / IL1B Protein

A DNA sequence encoding the Mus musculus (Mouse) IL-1 beta / IL1B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8893-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017031

Quantity Price
1 3211.16€ / 1mg
3 3021.38€ / 1mg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_032387.1
IL-1 beta / IL1B Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
16176
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IL1B
Recombinant Protein
E. coli
PIRQLHYRLR DEQQKSLVLS DPYELKALHL NGQNINQQVI FSMSFVQGEP SNDKIPVALG LKGKNLYLSC VMKDGTPTLQ LESVDPKQYP KKKMEKRFVF NKIEVKSKVE FESAEFPNWY ISTSQAEHKP VFLGNNSGQD IIDFTMESVS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers