missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Histone H3-like centromeric protein 1

A DNA sequence encoding the Mus musculus (Mouse) Histone H3-like centromeric protein A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9060-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017551

  • 770.00€ / 100µg
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001289058.1
Histone H3-like centromeric protein 1
Research Use Only
Cenpa
Recombinant Protein
Yeast
MGPRRKPQTP RRRPSSPAPG PSRQSSSVGS QTLRRRQKFM WLKEIKTLQK STDLLFRKKP FSMVVREICE KFSRGVDFWW QAQALLALQE AAEAFLIHLF EDAYLLSLHA GRVTLFPKDI QLTRRIRGFE GGLP
Greater than 90% as determined by SDS-PAGE.
12615
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers