missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse F-box / WD repeat-containing protein 10 Protein
A DNA sequence encoding the Mus musculus (Mouse) F-box/WD repeat-containing protein 10, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Name | F-box / WD repeat-containing protein 10 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | Fbxw10 |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16000331
|
enQuireBio™
QP7709-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16020331
|
enQuireBio™
QP7709-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16010331
|
enQuireBio™
QP7709-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16030331
|
enQuireBio™
QP7709-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16040331
|
enQuireBio™
QP7709-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16050331
|
enQuireBio™
QP7709-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
F-box / WD repeat-containing protein 10 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
Fbxw10 | |
Recombinant Protein | |
E. coli | |
VKWQYSSDKN KVKKSKDKEE EREETSLGDE HSRSTIQGHS LKDSVSSKQE FSKSRVHLKQ TKNLSSDDME TPVGEVSHPL QKLWKVPMTP DRFLLTISAL QQAHNSEEFA YPHRPRPQVI DAWGPSIPYP RKVLSLKGKS VQHAVDQLRS SNLPTGVRQT NIPLEIQKLQ PNLKKSLHSP RVQATVPQPS LIRPKVSDSL RGDEHLTSSI DGTMRRAGPL TSMQVIKPNR MLAPRGGTAT LSPKKERPRF YTTLDPLRMN TGFMLMTVKE EKEFAEAKMK EYEASVSTKE VDPGKASKAA WIRKIKGLPI DNFMKEGKTA APELGQNVFI | |
Greater than 90% as determined by SDS-PAGE. |