missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse E3 ubiquitin-protein ligase TRIM21 Protein
A DNA sequence encoding the Mus musculus (Mouse) E3 ubiquitin-protein ligase TRIM21, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7725-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
E3 ubiquitin-protein ligase TRIM21 Protein | |
Research Use Only | |
TRIM21 | |
Recombinant Protein | |
E. coli | |
MSPSTTSKMS LEKMWEEVTC SICLDPMVEP MSIECGHCFC KECIFEVGKN GGSSCPECRQ QFLLRNLRPN RHIANMVENL KQIAQNTKKS TQETHCMKHG EKLHLFCEED GQALCWVCAQ SGKHRDHTRV PIEEAAKVYQ EKIHVVLEKL RKGKELAEKM EMDLTMQRTD WKRNIDTQKS RIHAEFALQN SLLAQEEQRQ LQRLEKDQRE YLRLLGKKEA ELAEKNQALQ ELISELERRI RGSELELLQE VRIILERSGS WNLDTLDIDA PDLTSTCPVP GRKKMLRTCW VHITLDRNTA NSWLIISKDR RQVRMGDTHQ NVSDNKERFS NYPMVLGAQR FSSGKMYWEV DVTQKEAWDL GVCRDSVQRK GQFSLSPENG FWTIWLWQDS YEAGTSPQTT LHIQVPPCQI GIFVDYEAGV VSFYNITDHG SLIYTFSECV FAGPLRPFFN VGFNYSGGNA APLKLCPLKM | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |