missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CXCL16 / SR-PSOX Protein

A DNA sequence encoding the Mus musculus (Mouse) CXCL16 / SR-PSOX, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8659-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16025821

Quantity Price
1 3211.16€ / 1mg
3 3021.38€ / 1mg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_075647.3
CXCL16 / SR-PSOX Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
66102
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Cxcl16
Recombinant Protein
E. coli
NQGSVAGSCS CDRTISSGTQ IPQGTLDHIR KYLKAFHRCP FFIRFQLQSK SVCGGSQDQW VRELVDCFER KECGTGHGKS FHHQKHLPQA STQTPEAAEG TPSDTSTPAH SQSTQHSTLP SGALSLNKEH TQPWEMTTLP SGYGLEARPE AEANEKQQDD RQQEAPGAGA ST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers