missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD3e / CD3 epsilon Protein

A DNA sequence encoding the Mus musculus (Mouse) CD3e / CD3 epsilon, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9045-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017521

Quantity Price
1 725.49€ / 100µg
5 682.62€ / 100µg
Estimated Shipment: 19-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_031674.1
CD3e / CD3 epsilon Protein
100 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
12501
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CD3E
Recombinant Protein
Yeast
DAENIEYKVS ISGTSVELTC PLDSDENLKW EKNGQELPQK HDKHLVLQDF SEVEDSGYYV CYTPASNKNT YLYLKARVCE YCVEVD
Videos
SDS
Documents
Certificates
Special Offers

Special Offers