missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Cd163 Protein

A DNA sequence encoding the Mus musculus (Mouse) Cd163, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7641-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029970

  • 3405.00€ / 1mg
Estimated Shipment: 19-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_444324.2
Cd163 Protein
Research Use Only
Cd163
Recombinant Protein
E. coli
VVCQQLGCPT SIKALGWANS SAGSGYIWMD KVSCTGNESA LWDCKHDGWG KHNCTHEKDA GVTCSDGSNL EMRLVNSAGH RCLGRVEIKF QGKWGTVCDD NFSKDHASVI CKQLGCGSAI SFSGSAKLGA GSGPIWLDDL ACNGNESALW DCKHRGWGKH NCDHAEDVGV ICLEGADLSL RLVDGVSRCS GRLEVRFQGE WGTVCDDNWD LRDASVVCKQ LGCPTAISAI GRVNASEGSG QIWLDNISCE GHEATLWECK HQEWGKHYCH HREDAGVTCS
Greater than 90% as determined by SDS-PAGE.
93671
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers