missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse bFGF / FGF2 Protein

A DNA sequence encoding the Mus musculus (Mouse) bFGF / FGF2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8538-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035171

  • 3405.00€ / 1mg
Estimated Shipment: 28-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_032032.1
bFGF / FGF2 Protein
Research Use Only
FGF2
Recombinant Protein
E. coli
PALPEDGGAA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHV KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTEE CFFFERLESN NYNTYRSRKY SSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
Greater than 90% as determined by SDS-PAGE.
14173
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers