missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Adamts5 Protein

A DNA sequence encoding the Mus musculus (Mouse) Adamts5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8228-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023411

  • 3405.00€ / 1mg
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_035912.2
Adamts5 Protein
Research Use Only
Adamts5
Recombinant Protein
E. coli
SISRARQVEL LLVADSSMAR MYGRGLQHYL LTLASIANRL YSHASIENHI RLAVVKVVVL TDKDTSLEVS KNAATTLKNF CKWQHQHNQL GDDHEEHYDA AILFTREDLC GHHSCDTLGM ADVGTICSPE RSCAVIEDDG LHAAFTVAHE IGHLLGLSHD DSKFCEENFG TTEDKRLMSS ILTSIDASKP WSKCTSATIT EFLDDGHGNC LLDLPRKQIL GPEELPGQTY DATQQCNLTF GPEYSVCPGM DVCARLWCAV VRQGQMVCLT KKLPAVEGTP CGKGRVCLQG KCVDKTKKKY YSTSSHGNWG SWGPWGQCSR SCGGGVQFAY RHCNNPAPRN SGRYCTGKRA IYRSCSVTPC PPNGKSFRHE QCEAKNGYQS DAKGVKTFVE WVPKYAGVLP ADVCKLTCRA KGTGYYVVFS PKVTDGTECR PYSNSVCVRG RCVRTGCDGI IGSKLQYDKC GVCGGDNSSC TKIIGTFNKK SKGYTDVVRI PEGATHIKVR QFKAKDQTRF TAYLALKKKT GEYLINGKYM ISTSETIIDI NGTVMNYSGW SHRDDFLHGM GYSATKEILI VQILATDPTK ALDVRYSFFV PKKTTQKVNS VISHGSNKVG PHSTQLQWVT GPWLACSRTC DTGWHTRTVQ CQDGNRKLAK GCLLSQRPSA FKQCLLKKC
Greater than 90% as determined by SDS-PAGE.
23794
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers