missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL-1 alpha / IL1A / IL1F1 Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL-1 alpha / IL1A / IL1F1 Protein, was expressed in the hosts and tags indicated.
352.00€ - 3175.00€
Specifications
Accession Number | NP_001036222.1 |
---|---|
Gene ID (Entrez) | 700193 |
Name | IL-1 alpha / IL1A / IL1F1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091620
|
enQuireBio™
QP6213-YE-100UG |
100 μg |
1100.00€
100µg |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
16031630
|
enQuireBio™
QP6213-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
16001630
|
enQuireBio™
QP6213-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16021630
|
enQuireBio™
QP6213-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16041630
|
enQuireBio™
QP6213-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16011630
|
enQuireBio™
QP6213-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001036222.1 | |
IL-1 alpha / IL1A / IL1F1 Protein | |
Not determined. | |
Full Length | |
Macaque | |
His | |
Tris-based buffer, 50% glycerol |
700193 | |
Research Use Only | |
IL1A | |
Recombinant Protein | |
Yeast | |
SAPFSFLSNM TYHFIRIIKH EFILNDTLNQ TIIRANDQHL TAAAIHNLDE AVKFDMGAYT SSKDDTKVPV ILRISKTQLY VSAQDEDQPV LLKEMPEINK TITGSETNFL FFWETHGTKN YFISVAHPNL FIATKHDNWV CLAKGLPSIT DFQILENQA | |
Greater than 90% as determined by SDS-PAGE. |