missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant M. tuberculosis Immunogenic protein MPT64 Protein

A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Immunogenic protein MPT64, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9654-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028561

  • 1100.00€ / 100µg
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

WP_003409954.1
100 μg
Not determined.
Full Length
M. tuberculosis
His
Tris-based buffer, 50% glycerol
M. tuberculosis Immunogenic protein MPT64 Protein
Research Use Only
mpt64
Recombinant Protein
Yeast
APKTYCEELK GTDTGQACQI QMSDPAYNIN ISLPSYYPDQ KSLENYIAQT RDKFLSAATS STPREAPYEL NITSATYQSA IPPRGTQAVV LKVYQNAGGT HPTTTYKAFD WDQAYRKPIT YDTLWQADTD PLPVVFPIVQ GELSKQTGQQ VSIAPNAGLD PVNYQNFAVT NDGVIFFFNP GELLPEAAGP TQVLVPRSAI DSMLA
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions