missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza B (strain B / Lee / 1940) NucleoProtein
A DNA sequence encoding the Influenza B virus (strain B/Lee/1940) Influenza B (strain B/Lee/1940) Nucleoprotein, was expressed in the hosts and tags indicated.
352.00€ - 3175.00€
Specifications
Accession Number | NP_056661.1 |
---|---|
Gene ID (Entrez) | 26824002 |
Name | Influenza B (strain B / Lee / 1940) NucleoProtein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028420
|
enQuireBio™
QP7357-YE-100UG |
100 μg |
1100.00€
100µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16068420
|
enQuireBio™
QP7357-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16038420
|
enQuireBio™
QP7357-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16058420
|
enQuireBio™
QP7357-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16078420
|
enQuireBio™
QP7357-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16048420
|
enQuireBio™
QP7357-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_056661.1 | |
Influenza B (strain B / Lee / 1940) NucleoProtein | |
Not determined. | |
Full Length | |
Influenza | |
His | |
Tris-based buffer, 50% glycerol |
26824002 | |
Research Use Only | |
NP | |
Recombinant Protein | |
Yeast | |
MSNMDIDSIN TGTIDKTPEE LTPGTSGATR PIIKPATLAP PSNKRTRNPS PERTTTSSET DIGRKIQKKQ TPTEIKKSVY KMVVKLGEFY NQMMVKAGLN DDMERNLIQN AQAVERILLA ATDDKKTEYQ KKRNARDVKE GKEEIDHNKT GGTFYKMVRD DKTIYFSPIK ITFLKEEVKT MYKTTMGSDG FSGLNHIMIG HSQMNDVCFQ RSKGLKRVGL DPSLISTFAG STLPRRSGTT GVAIKGGGTL VDEAIRFIGR AMADRGLLRD IKAKTAYEKI LLNLKNKCSA PQQKALVDQV IGSRNPGIAD IEDLTLLARS MVVVRPSVAS KVVLPISIYA KIPQLGFNTE EYSMVGYEAM ALYNMATPVS ILRMGDDAKD KSQLFFMSCF GAAYEDLRVL SALTGTEFKP RSALKCKGFH VPAKEQVEGM GAALMSIKLQ FWAPMTRSGG NEVSGEGGSG QISCSPVFAV ERPIALSKQA VRRMLSMNVE GRDADVKGNL LKMMNDSMAK KTSGNAFIGK KMFQISDKNK VNPIEIPIKQ TIPNFFFGRD TAEDYDDLDY | |
Greater than 90% as determined by SDS-PAGE. |