missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza A H3N2 (Hong Kong / 1 / 1968) Nucleoprotein / NP Protein

A DNA sequence encoding the Influenza A virus (strain A/Hong Kong/1/1968 H3N2) Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein / NP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7093-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16027370

Quantity Price
1 584.93€ / 50µg
6 550.25€ / 50µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

Influenza A H3N2 (Hong Kong / 1 / 1968) Nucleoprotein / NP Protein
50 μg
Not determined.
Full Length
Influenza
His-SUMO
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
NP
Recombinant Protein
E. coli
MASQGTKRSY EQMETDGERQ NATEIRASVG KMIDGIGRFY IQMCTELKLS DYEGRLIQNS LTIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYKRV DGKWMRELVL YDKEEIRRIW RQANNGDDAT AGLTHMMIWH SNLNDTTYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTMV MELIRMIKRG INDRNFWRGE NGRKTRSAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGPAVASGYD FEKEGYSLVG IDPFKLLQNS QVYSLIRPNE NPAHKSQLVW MACNSAAFED LRVLSFIRGT KVSPRGKLST RGVQIASNEN MDAMESSTLE LRSRYWAIRT RSGGNTNQQR ASAGQISVQP AFSVQRNLPF DKPTIMAAFT GNTEGRTSDM RAEIIRMMEG AKPEEMSFQG RGVFELSDEK AANPIVPSFD MSNEGSYFFG DNAEEYDN
Videos
SDS
Documents
Certificates
Special Offers

Special Offers