missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zinc finger protein 165 Protein

A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 165, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6903-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016660

Quantity Price
1 2248.90€ / 1mg
3 2115.77€ / 1mg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_003438.1
Zinc finger protein 165 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
7718
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ZNF165
Recombinant Protein
E. coli
MATEPKKAAA QNSPEDEGLL IVKIEEEEFI HGQDTCLQRS ELLKQELCRQ LFRQFCYQDS PGPREALSRL RELCCQWLKP EIHTKEQILE LLVLEQFLTI LPGDLQAWVH EHYPESGEEA VTILEDLERG TDEAVLQVQA HEHGQEIFQK KVSPPGPALN VKLQPVETKA HFDSSEPQLL WDCDNESENS RSMPKLEIFE KIESQRIISG RISGYISEAS GESQDICKSA GRVKRQWEKE SGESQRLSSA QDEGFGKILT HKNTVRGEII SHDGCERRLN LNSNEFTHQK SCKHGTCDQS FKWNSDFINH QIIYAGEKNH QYGKSFKSPK LAKHAAVFSG DKTHQCNECG KAFRHSSKLA RHQRIHTGER CYECNECGKS FAESSDLTRH RRIHTGERPF GCKECGRAFN LNSHLIRHQR IHTREKPYEC SECGKTFRVS SHLIRHFRIH TGEKPYECSE CGRAFSQSSN LSQHQRIHMR ENLLM
Videos
SDS
Documents
Certificates
Special Offers

Special Offers