missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VILIP-1 / VSNL1 Protein

A DNA sequence encoding the Homo sapiens (Human) VILIP-1 / VSNL1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6883-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016540

  • 2385.00€ / 1mg
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_003376.2
VILIP-1 / VSNL1 Protein
Research Use Only
VSNL1
Recombinant Protein
E. coli
GKQNSKLAPE VMEDLVKSTE FNEHELKQWY KGFLKDCPSG RLNLEEFQQL YVKFFPYGDA SKFAQHAFRT FDKNGDGTID FREFICALSI TSRGSFEQKL NWAFNMYDLD GDGKITRVEM LEIIEAIYKM VGTVIMMKMN EDGLTPEQRV DKIFSKMDKN KDDQITLDEF KEAAKSDPSI VLLLQCDIQK
Greater than 80% as determined by SDS-PAGE.
7447
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers