missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VGLL3 Protein
A DNA sequence encoding the Homo sapiens (Human) VGLL3, was expressed in the hosts and tags indicated.
202.00€ - 2155.00€
Specifications
Accession Number | NP_001307422.1 |
---|---|
Gene ID (Entrez) | 389136 |
Name | VGLL3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16018361
|
enQuireBio™
QP9465-YE-100UG |
100 μg |
645.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16058361
|
enQuireBio™
QP9465-YE-500UG |
500 μg |
1740.00€
500µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16028361
|
enQuireBio™
QP9465-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16048361
|
enQuireBio™
QP9465-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16068361
|
enQuireBio™
QP9465-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16038361
|
enQuireBio™
QP9465-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001307422.1 | |
VGLL3 Protein | |
Not determined. | |
Full-Length (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
389136 | |
Research Use Only | |
VGLL3 | |
Recombinant Protein | |
Yeast | |
MSCAEVMYHP QPYGASQYLP NPMAATTCPT AYYQPAPQPG QQKKLAVFSK MQDSLEVTLP SKQEEEDEEE EEEEKDQPAE MEYLNSRCVL FTYFQGDIGS VVDEHFSRAL GQAITLHPES AISKSKMGLT PLWRDSSALS SQRNSFPTSF WTSSYQPPPA PCLGGVHPDF QVTGPPGTFS AADPSPWPGH NLHQTGPAPP PAVSESWPYP LTSQVSPSYS HMHDVYMRHH HPHAHMHHRH RHHHHHHHPP AGSALDPSYG PLLMPSVHAA RIPAPQCDIT KTEPTTVTSA TSAWAGAFHG TVDIVPSVGF DTGWSAMARS | |
Greater than 90% as determined by SDS-PAGE. |