missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Unconventional myosin-XIX Protein

A DNA sequence encoding the Homo sapiens (Human) Unconventional myosin-XIX, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7873-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16041301

Quantity Price
1 493.03€ / 100µg
6 463.44€ / 100µg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001028752.1
Unconventional myosin-XIX Protein
100 μg
Not determined.
Full Length of Isoform 4
Human
His-SUMO
Tris-based buffer, 50% glycerol
80179
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MYO19
Recombinant Protein
E. coli
LQQVNGHNPG SDGQAREYLR EDLQEFLGGE VLLYKLDDLT RVNPVTLETV LRCLQARYMA DTFYTNAGCT LVALNPFKPV PQLYSPELMR EYHAAPQPQK LKPHVFTVGE QTYRNVKSLI EPVNQSIVVS GESGAGKTWT SRCLMKFYAV VATSPASWES HKIAERIEQR ILNSNPVMEA FGNACTLRNN NSSRFGKFIQ LQLNRAQQMT GAAVQTYLLE KTRVACQASS ERNFHIFYQI CKGASEDERL QWHLPEGAAF SWLPNPERSL EEDCFEVTRE AMLHLGIDTP TQNNIFKVLA GLLHLGNIQF AASEDEAQPC QPMDDAKYSV RTAASLLGLP EDVLLEMVQI RTIRAGRQQQ VFRKPCARAE CDTRRDCLAK LIYARLFDWL VSVINSSICA DTDSWTTFIG LLDVYGFESF PDNSLEQLCI NYANEKLQLE ACGLVETIHI SAAGFPIRVS HRNFVERYKL LRRLHPCTSS GPDSPYPAKG LPEWCPHSEE ATLEPLIQDI LHTLPVLTQA AAITGDSAEA MPAPMHCGRT KVFMTDSMLE LLECGRARVL EQCARCIQGG WRRHRHREQE RQWRAVMLIQ AAIRSWLTRK HIQRLHAAAT VIKRAWQKWR IRMACLAAKE LDGVEEKHFS QAPCSLSTSP LQTRLLEAII RLWPLGLVLA NTAMGVGSFQ RKLVVWACLQ LPRGSPSSYT VQTAQDQAGV TSIRALPQGS IKFHCRKSPL RYADICPEPS PYSITGFNQI LLERHRLIHV TSSAFTGLG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers