missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ubiquitin-like-conjugating enzyme ATG10 Protein

A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-like-conjugating enzyme ATG10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8050-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022371

  • 493.03€ / 100µg

Estimated Shipment: 13-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_001124500.1
Ubiquitin-like-conjugating enzyme ATG10 Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
83734
Greater than 80% as determined by SDS-PAGE.
Research Use Only
ATG10
Recombinant Protein
E. coli
MEEDEFIGEK TFQRYCAEFI KHSQQIGDSW EWRPSKDCSD GYMCKIHFQI KNGSVMSHLG ASTHGQTCLP MEEAFELPLD DCEVIETAAA SEVIKYEYHV LYSCSYQVPV LYFRASFLDG RPLTLKDIWE GVHECYKMRL LQGPWDTITQ QEHPILGQPF FVLHPCKTNE FMTPVLKNSQ KINKNVNYIT SWLSIVGPVV GLNLPLSYAK ATSQDERNVP
Videos
SDS
Documents
Certificates
Special Offers

Special Offers