missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transmembrane protein 44 Protein

A DNA sequence encoding the Homo sapiens (Human) Transmembrane protein 44, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6806-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015980

Quantity Price
1 493.03€ / 100µg
6 463.44€ / 100µg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001011655.1
Transmembrane protein 44 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
93109
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TMEM44
Recombinant Protein
E. coli
MKSKMRQALG FAKEARESPD TQALLTCAEK EEENQENLDW VPLTTLSHCK SLRTMTAISR YMELTIEPVQ QAGCSATRLP GDGQTSAGDA SLQDPPSYPP VQVIRARVSS GSSSEVSSIN SDLEQKYWEA LNSEQWDPED VNLEGSKENV ELLGSQVHQD SVRTAHLSDD D
Videos
SDS
Documents
Certificates
Special Offers

Special Offers