missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcription elongation factor A protein 1

A DNA sequence encoding the Homo sapiens (Human) Transcription elongation factor A protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7582-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029550

  • 2385.00€ / 1mg
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_003186.1
Transcription elongation factor A protein 1
Research Use Only
TCEA2
Recombinant Protein
E. coli
MMGKEEEIAR IARRLDKMVT KKSAEGAMDL LRELKAMPIT LHLLQSTRVG MSVNALRKQS SDEEVIALAK SLIKSWKKLL DASDAKARER GRGMPLPTSS RDASEAPDPS RKRPELPRAP STPRITTFPP VPVTCDAVRN KCREMLTAAL QTDHDHVAIG ADCERLSAQI EECIFRDVGN TDMKYKNRVR SRISNLKDAK NPDLRRNVLC GAITPQQIAV MTSEEMASDE LKEIRKAMTK EAIREHQMAR TGGTQTDLFT CGKCRKKNCT YTQVQTRSSD EPMTTFVVCN ECGNRWKFC
Greater than 90% as determined by SDS-PAGE.
6919
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers