missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcription activator BRG1 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcription activator BRG1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9406-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001122316.1 | |
Transcription activator BRG1 Protein | |
Research Use Only | |
SMARCA4 | |
Recombinant Protein | |
Yeast | |
EVDARHIIEN AKQDVDDEYG VSQALARGLQ SYYAVAHAVT ERVDKQSALM VNGVLKQYQI KGLEWLVSLY NNNLNGILAD EMGLGKTIQT IALITYLMEH KRINGPFLII VPLSTLSNWA YEFDKWAPSV VKVSYKGSPA ARRAFVPQLR SGKFNVLLTT YEYIIKDKHI LAKIRWKYMI VDEGHRMKNH HCKLTQVLNT HYVAPRRLLL TGTPLQNKLP ELWALLNFLL PTIFKSCSTF EQWFNAPFAM TGEKVDLNEE ETILIIRRLH KVLRPFLLRR LKKEVEAQLP EKVEYVIKCD MSALQRVLYR HMQAKGVLLT DGSEKDKKGK GGTKTLMNTI MQLRKICNHP YMFQHIEESF SEHLGFTGGI VQGLDLYRAS GKFELLDRIL PKLRATNHKV LLFCQMTSLM TIMEDYFAYR GFKYLRLDGT TKAEDRGMLL KTFNEPGSEY FIFLLSTRAG GLGLNLQSAD TVIIFDSDWN PHQDLQAQDR AHRIGQQNEV RVLRLCTVNS VEEKILAAAK YKLNVDQKVI QAGMFDQKSS SHERRAF | |
Greater than 90% as determined by SDS-PAGE. |
6597 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |