missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Talin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Talin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8498-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024961

Quantity Price
1 1643.42€ / 500µg
3 1544.82€ / 500µg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_006280.3
Talin-1 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
7094
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TLN1
Recombinant Protein
Yeast
MLDGTVKTIM VDDSKTVTDM LMTICARIGI TNHDEYSLVR ELMEEKKEEI TGTLRKDKTL LRDEKKMEKL KQKLHTDDEL NWLDHGRTLR EQGVEEHETL LLRRKFFYSD QNVDSRDPVQ LNLLYVQARD DILNGSHPVS FDKACEFAGF QCQIQFGPHN EQKHKAGFLD LKDFLPKEYV KQKGERKIFQ AHKNCGQMSE IEAKVRYVKL ARSLKTYGVS FFLVKEKMKG KNKLVPRLLG ITKECVMRVD EKTKEVIQEW NLTNIKRWAA SPKSFTLDFG DYQDGYYSVQ TTEGEQIAQL IAGYIDII
Videos
SDS
Documents
Certificates
Special Offers

Special Offers