missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Syndecan-4 / SDC4 Protein
A DNA sequence encoding the Homo sapiens (Human) Syndecan-4 / SDC4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7123-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_002990.2 | |
Syndecan-4 / SDC4 Protein | |
Research Use Only | |
SDC4 | |
Recombinant Protein | |
E. coli | |
ESIRETEVID PQDLLEGRYF SGALPDDEDV VGPGQESDDF ELSGSGDLDD LEDSMIGPEV VHPLVPLDNH IPERAGSGSQ VPTEPKKLEE NEVIPKRISP VEESEDVSNK VSMSSTVQGS NIFERTE | |
Greater than 90% as determined by SDS-PAGE. |
6385 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |