missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Synaptotagmin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Synaptotagmin-1, was expressed in the hosts and tags indicated.

160.00€ - 2180.00€

Specifications

Accession Number NP_001129277.1
Gene ID (Entrez) 6857
Name Synaptotagmin-1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16095600
View Documents Promotion Details
enQuireBio™
QP6757-EC-100UG
100 μg
476.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 22-11-2023
to see stock.
Add to basket
 
16015610
View Documents Promotion Details
enQuireBio™
QP6757-EC-1MG
1 mg
2180.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 22-11-2023
to see stock.
Add to basket
 
16005610
View Documents Promotion Details
enQuireBio™
QP6757-EC-10UG
10 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
16025610
View Documents Promotion Details
enQuireBio™
QP6757-EC-200UG
200 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
16035610
View Documents Promotion Details
enQuireBio™
QP6757-EC-500UG
500 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
16045610
View Documents Promotion Details
enQuireBio™
QP6757-EC-50UG
50 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001129277.1
Synaptotagmin-1 Protein
Not determined.
Cytoplasmic Domain
Human
His
Tris-based buffer, 50% glycerol
6857
Research Use Only
SYT1
Recombinant Protein
E. coli
KNAINMKDVK DLGKTMKDQA LKDDDAETGL TDGEEKEEPK EEEKLGKLQY SLDYDFQNNQ LLVGIIQAAE LPALDMGGTS DPYVKVFLLP DKKKKFETKV HRKTLNPVFN EQFTFKVPYS ELGGKTLVMA VYDFDRFSKH DIIGEFKVPM NTVDFGHVTE EWRDLQSAEK EEQEKLGDIC FSLRYVPTAG KLTVVILEAK NLKKMDVGGL SDPYVKIHLM QNGKRLKKKK TTIKKNTLNP YYNESFSFEV PFEQIQKVQV VVTVLDYDKI GKNDAIGKVF VGYNSTGAEL RHWSDMLANP RRPIAQWHTL QVEEEVDA
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications