missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Synaptosomal-associated protein 29 Protein

A DNA sequence encoding the Homo sapiens (Human) Synaptosomal-associated protein 29, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6703-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035240

  • 530.00€ / 100µg
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_004773.1
Synaptosomal-associated protein 29 Protein
Research Use Only
SNAP29
Recombinant Protein
E. coli
MSAYPKSYNP FDDDGEDEGA RPAPWRDARD LPDGPDAPAD RQQYLRQEVL RRAEATAAST SRSLALMYES EKVGVASSEE LARQRGVLER TEKMVDKMDQ DLKISQKHIN SIKSVFGGLV NYFKSKPVET PPEQNGTLTS QPNNRLKEAI STSKEQEAKY QASHPNLRKL DDTDPVPRGA GSAMSTDAYP KNPHLRAYHQ KIDSNLDELS MGLGRLKDIA LGMQTEIEEQ DDILDRLTTK VDKLDVNIKS TERKVRQL
Greater than 80% as determined by SDS-PAGE.
9342
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers