missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SUB1 Protein

A DNA sequence encoding the Homo sapiens (Human) SUB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6745-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035530

Quantity Price
1 2248.90€ / 1mg
3 2115.77€ / 1mg
Estimated Shipment: 19-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_006704.3
SUB1 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
10923
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SUB1
Recombinant Protein
E. coli
PKSKELVSSS SSGSDSDSEV DKKLKRKKQV APEKPVKKQK TGETSRALSS SKQSSSSRDD NMFQIGKMRY VSVRDFKGKV LIDIREYWMD PEGEMKPGRK GISLNPEQWS QLKEQISDID DAVRKL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers