missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STOM / EPB72 Protein

A DNA sequence encoding the Homo sapiens (Human) STOM / EPB72, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6740-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015500

Quantity Price
1 725.49€ / 100µg
5 682.62€ / 100µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_004090.4
STOM / EPB72 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
2040
Greater than 90% as determined by SDS-PAGE.
Research Use Only
STOM
Recombinant Protein
E. coli
MAEKRHTRDS EAQRLPDSFK DSPSKGLGPC GWILVAFSFL FTVITFPISI WMCIKIIKEY ERAIIFRLGR ILQGGAKGPG LFFILPCTDS FIKVDMRTIS FDIPPQEILT KDSVTISVDG VVYYRVQNAT LAVANITNAD SATRLLAQTT LRNVLGTKNL SQILSDREEI AHNMQSTLDD ATDAWGIKVE RVEIKDVKLP VQLQRAMAAE AEASREARAK VIAAEGEMNA SRALKEASMV ITESPAALQL RYLQTLTTIA AEKNSTIVFP LPIDMLQGII GAKHSHLG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers