missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Stathmin-4 Protein

A DNA sequence encoding the Homo sapiens (Human) Stathmin-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8172-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16033051

  • 530.00€ / 100µg
Estimated Shipment: 28-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001269982.1
Stathmin-4 Protein
Research Use Only
STMN4
Recombinant Protein
E. coli
MTLAAYKEKM KELPLVSLFC SCFLADPLNK SSYKYEGWCG RQCRRKDESQ RKDSADWRER RAQADTVDLN WCVISDMEVI ELNKCTSGQS FEVILKPPSF DGVPEFNASL PRRRDPSLEE IQKKLEAAEE RRKYQEAELL KHLAEKREHE REVIQKAIEE NNNFIKMAKE KLAQKMESNK ENREAHLAAM LERLQEKDKH AEEVRKNKEL KEEASR
Greater than 80% as determined by SDS-PAGE.
81551
100 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers