missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Src kinase-associated phosphoprotein 1

A DNA sequence encoding the Homo sapiens (Human) Src kinase-associated phosphoprotein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7785-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16020781

Quantity Price
1 493.03€ / 100µg
6 463.44€ / 100µg
Estimated Shipment: 19-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001068567.1
Src kinase-associated phosphoprotein 1
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
8631
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SKAP1
Recombinant Protein
E. coli
MQAAALPEEI RWLLEDAEEF LAEGLRNENL SAVARDHRDH ILRGFQQIKA RYYWDFQPQG GDIGQDSSDD NHSGTLGLSL TSDAPFLSDY QDEGMEDIVK GAQELDNVIK QGYLEKKSKD HSFFGSEWQK RWCVVSRGLF YYYANEKSKQ PKGTFLIKGY GVRMAPHLRR DSKKESCFEL TSQDRRSYEF TATSPAEARD WVDQISFLLK DLSSLTIPYE EDEEEEEKEE TYDDIDGFDS PSCGSQCRPT ILPGSVGIKE PTEEKEEEDI YEVLPDEEHD LEEDESGTRR KGVDYASYYQ GLWDCHGDQP DELSFQRGDL IRILSKEYNM YGWWVGELNS LVGIVPKEYL TTAFEVEE
Videos
SDS
Documents
Certificates
Special Offers

Special Offers