missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Sclerostin domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) Sclerostin domain-containing protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6717-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015350

  • 530.00€ / 100µg
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_056279.1
Sclerostin domain-containing protein 1
Research Use Only
SOSTDC1
Recombinant Protein
E. coli
FKNDATEILY SHVVKPVPAH PSSNSTLNQA RNGGRHFSNT GLDRNTRVQV GCRELRSTKY ISDGQCTSIS PLKELVCAGE CLPLPVLPNW IGGGYGTKYW SRRSSQEWRC VNDKTRTQRI QLQCQDGSTR TYKITVVTAC KCKRYTRQHN ESSHNFESMS PAKPVQHHRE RKRASKSSKH SMS
Greater than 90% as determined by SDS-PAGE.
25928
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers