missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SART3 Protein

A DNA sequence encoding the Homo sapiens (Human) SART3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7616-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16039810

  • 2385.00€ / 1mg
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_055521.1
SART3 Protein
Research Use Only
SART3
Recombinant Protein
E. coli
QRKRARAEKK ALKKKKKIRG PEKRGADEDD EKEWGDDEEE QPSKRRRVEN SIPAAGETQN VEVAAGPAGK CAAVDVEPPS KQKEKAASLK RDMPKVLHDS SKDSITVFVS NLPYSMQEPD TKLRPLFEAC GEVVQIRPIF SNRGDFRGYC YVEFKEEKSA LQALEMDRKS VEGRPMFVSP CVDKSKNPDF KVFRYSTSLE KHKLFISGLP FSCTKEELEE ICKAHGTVKD LRLVTNRAGK PKGLAYVEYE NESQASQAVM KMDGMTIKEN IIKVAISNPP QRKVPEKPET RKAPGGPMLL P
Greater than 90% as determined by SDS-PAGE.
9733
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers