missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ropporin-1B Protein
A DNA sequence encoding the Homo sapiens (Human) Ropporin-1B, was expressed in the hosts and tags indicated.
202.00€ - 2155.00€
Specifications
Accession Number | NP_001012337.1 |
---|---|
Gene ID (Entrez) | 152015 |
Name | Ropporin-1B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024560
|
enQuireBio™
QP6614-YE-100UG |
100 μg |
645.00€
100µg |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
16064560
|
enQuireBio™
QP6614-YE-500UG |
500 μg |
1740.00€
500µg |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
16034560
|
enQuireBio™
QP6614-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16054560
|
enQuireBio™
QP6614-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16074560
|
enQuireBio™
QP6614-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16044560
|
enQuireBio™
QP6614-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001012337.1 | |
Ropporin-1B Protein | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 2 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
152015 | |
Research Use Only | |
ROPN1B | |
Recombinant Protein | |
Yeast | |
MWKVVNLPTD LFNSVMNVGR FTEEIEWLKF LALACSALGV TITKTLKIVC EVLSCDHNGG LPRIPFSTFQ FLYTYIAEVD GEICASHVSR MLNYIEQEVI GPDGLITVND FTQNPRVWLE | |
Greater than 90% as determined by SDS-PAGE. |