missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Regulator of G-protein signaling 7 Protein
A DNA sequence encoding the Homo sapiens (Human) Regulator of G-protein signaling 7, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001269702.1 |
---|---|
Gene ID (Entrez) | 6000 |
Name | Regulator of G-protein signaling 7 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024500
|
enQuireBio™
QP6606-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16044500
|
enQuireBio™
QP6606-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16034500
|
enQuireBio™
QP6606-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16054500
|
enQuireBio™
QP6606-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16064500
|
enQuireBio™
QP6606-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16074500
|
enQuireBio™
QP6606-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001269702.1 | |
Regulator of G-protein signaling 7 Protein | |
Not determined. | |
Full Length of Isoform 5 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6000 | |
Research Use Only | |
RGS7 | |
Recombinant Protein | |
E. coli | |
MAQGNNYGQT SNGVADESPN MLVYRKMEDV IARMQDEKNG IPIRTVKSFL SKIPSVFSGS DIVQWLIKNL TIEDPVEALH LGTLMAAHGY FFPISDHVLT LKDDGTFYRF QTPYFWPSNC WEPENTDYAV YLCKRTMQNK ARLELADYEA ESLARLQRAF ARKWEFIFMQ AEAQAKVDKK RDKIERKILD SQERAFWDVH RPVPGCVNTT EVDIKKSSRM RNPHKTRKSV YGLQNDIRSH SPTHTPTPET KPPTEDELQQ QIKYWQIQLD RHRLKMSKVA DSLLSYTEQY LEYDPFLLPP DPSNPWLSDD TTFWELEASK EPSQQRVKRW GFGMDEALKD PVGREQFLKF LESEFSSENL RFWLAVEDLK KRPIKEVPSR VQEIWQEFLA PGAPSAINLD SKSYDKTTQN VKEPGRYTFE DAQEHIYKLM KSDSYPRFIR SSAYQELLQA KKKSGNSMDR RTSFEKFAQN VGKSLTSKRL TSLAQSY | |
Greater than 90% as determined by SDS-PAGE. |