missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human REG3A / HIP Protein

A DNA sequence encoding the Homo sapiens (Human) REG3A / HIP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6593-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024400

Quantity Price
1 2248.90€ / 1mg
3 2115.77€ / 1mg
Estimated Shipment: 14-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_002571.1
REG3A / HIP Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
5068
Greater than 90% as determined by SDS-PAGE.
Research Use Only
REG3A
Recombinant Protein
E. coli
EEPQRELPSA RIRCPKGSKA YGSHCYALFL SPKSWTDADL ACQKRPSGNL VSVLSGAEGS FVSSLVKSIG NSYSYVWIGL HDPTQGTEPN GEGWEWSSSD VMNYFAWERN PSTISSPGHC ASLSRSTAFL RWKDYNCNVR LPYVCKFTD
Videos
SDS
Documents
Certificates
Special Offers

Special Offers