missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Recoverin Protein

A DNA sequence encoding the Homo sapiens (Human) Recoverin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6591-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024390

Quantity Price
1 2497.57€ / 500µg
3 2349.71€ / 500µg
Estimated Shipment: 14-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_002894.1
Recoverin Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
5957
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RCVRN
Recombinant Protein
Yeast
GNSKSGALSK EILEELQLNT KFSEEELCSW YQSFLKDCPT GRITQQQFQS IYAKFFPDTD PKAYAQHVFR SFDSNLDGTL DFKEYVIALH MTTAGKTNQK LEWAFSLYDV DGNGTISKNE VLEIVMAIFK MITPEDVKLL PDDENTPEKR AEKIWKYFGK NDDDKLTEKE FIEGTLANKE ILRLIQFEPQ KVKEKMKNA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers