missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RBMY1A1 Protein

A DNA sequence encoding the Homo sapiens (Human) RBMY1A1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7035-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037130

Quantity Price
1 546.01€ / 100µg
6 513.42€ / 100µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001307873.1
RBMY1A1 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
378949
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RBMY1A1
Recombinant Protein
E. coli
MVEADHPGKL FIGGLNRETN EKMLKAVFGK HGPISEVLLI KDRTSKSRGF AFITFENPAD AKNAAKDMNG KSLHGKAIKV EQAKKPSFQS GGRRRPPASS RNRSPSGSLR SARGSRGGTR GWLPSHEGHL DDGGYTPDLK MSYSRGLIPV KRGPSSRSGG PPPKKSAPSA VARSNSWMGS QGPMSQRREN YGVPPRRATI SSWRNDRMST RHDGYATNDG NHPSCQETRD YAPPSRGYAY RDNGHSNRDE HSSRGYRNHR SSRETRDYAP PSRGHAYRDY GHSRRDESYS RGYRNRRSSR ETREYAPPSR GHGYRDYGHS RRHESYSRGY RNHPSSRETR DYAPPHRDYA YRDYGHSSWD EHSSRGYSYH DGYGEALGRD HSEHLSGSSY RDALQRYGTS HGAPPARGPR MSYGGSTCHA YSNTRDRYGR SWESYSSCGD FHYCDREHVC RKDQRNPPS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers