missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ras-related protein Rab-5A Protein

A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-5A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6571-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024250

  • 2248.90€ / 1mg

Estimated Shipment: 13-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_001278977.1
Ras-related protein Rab-5A Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
5868
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RAB5A
Recombinant Protein
E. coli
MASRGATRPN GPNTGNKICQ FKLVLLGESA VGKSSLVLRF VKGQFHEFQE STIGAAFLTQ TVCLDDTTVK FEIWDTAGQE RYHSLAPMYY RGAQAAIVVY DITNEESFAR AKNWVKELQR QASPNIVIAL SGNKADLANK RAVDFQEAQS YADDNSLLFM ETSAKTSMNV NEIFMAIAKK LPKNEPQNPG ANSARGRGVD LTEPTQPTRN QCCSN
Videos
SDS
Documents
Certificates
Special Offers

Special Offers