missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-23 Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-23, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8247-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001265595.1 | |
Ras-related protein Rab-23 Protein | |
Research Use Only | |
RAB23 | |
Recombinant Protein | |
E. coli | |
MLEEDMEVAI KMVVVGNGAV GKSSMIQRYC KGIFTKDYKK TIGVDFLERQ IQVNDEDVRL MLWDTAGQEE FDAITKAYYR GAQACVLVFS TTDRESFEAV SSWREKVVAE VGDIPTVLVQ NKIDLLDDSC IKNEEAEALA KRLKLRFYRT SVKEDLNVNE VFKYLAEKYL QKLKQQIAED PELTHSSSNK IGVFNTSGGS HSGQNSGTLN GGDVINLRPN KQRTKKNRNP FSSC | |
Greater than 80% as determined by SDS-PAGE. |
51715 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |