missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pyridoxal phosphate phosphatase Protein
A DNA sequence encoding the Homo sapiens (Human) Pyridoxal phosphate phosphatase, was expressed in the hosts and tags indicated.
655.00€ - 1815.00€
Specifications
Accession Number | NP_064711.1 |
---|---|
Gene ID (Entrez) | 57026 |
Name | Pyridoxal phosphate phosphatase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028861
|
enQuireBio™
QP9859-BA-20UG |
20 μg |
655.00€
20µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16018861
|
enQuireBio™
QP9859-BA-100UG |
100 μg |
1815.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16038861
|
enQuireBio™
QP9859-BA-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_064711.1 | |
Pyridoxal phosphate phosphatase Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
57026 | |
Research Use Only | |
Pdxp | |
Recombinant Protein | |
Baculovirus | |
MARCERLRGA ALRDVLGRAQ GVLFDCDGVL WNGERAVPGA PELLERLARA GKAALFVSNN SRRARPELAL RFARLGFGGL RAEQLFSSAL CAARLLRQRL PGPPDAPGAV FVLGGEGLRA ELRAAGLRLA GDPSAGDGAA PRVRAVLVGY DEHFSFAKLR EACAHLRDPE CLLVATDRDP WHPLSDGSRT PGTGSLAAAV ETASGRQALV VGKPSPYMFE CITENFSIDP ARTLMVGDRL ETDILFGHRC GMTTVLTLTG VSRLEEAQAY LAAGQHDLVP HYYVESIADL TEGLED | |
Greater than 90% as determined by SDS-PAGE. |