missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein prune homolog Protein

A DNA sequence encoding the Homo sapiens (Human) Protein prune homolog, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7790-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030821

  • 2385.00€ / 1mg
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001290158.1
Protein prune homolog Protein
Research Use Only
PRUNE
Recombinant Protein
E. coli
MLRKDQKTIY RQGVKVAISA IYMDLEICEV LERSHSPPLK LTPASSTHPN LHAYLQGNTQ VSRKKLLPLL QEALSAYFDS MKIPSGQPET ADVSREQVDK ELDRASNSLI SGLSQDEEDP PLPPTPMNSL VDECPLDQGL PKLSAEAVFE KCSQISLSQS TTASLSKK
Greater than 90% as determined by SDS-PAGE.
58497
1 mg
Not determined.
Full Length of Isoform 6
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers