missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein phosphatase 1 regulatory subunit 1B Protein
A DNA sequence encoding the Homo sapiens (Human) Protein phosphatase 1 regulatory subunit 1B, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_001229393.1 |
---|---|
Gene ID (Entrez) | 84152 |
Name | Protein phosphatase 1 regulatory subunit 1B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16012981
|
enQuireBio™
QP8161-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16032981
|
enQuireBio™
QP8161-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16022981
|
enQuireBio™
QP8161-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16042981
|
enQuireBio™
QP8161-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16052981
|
enQuireBio™
QP8161-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16062981
|
enQuireBio™
QP8161-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001229393.1 | |
Protein phosphatase 1 regulatory subunit 1B Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
84152 | |
Research Use Only | |
PPP1R1B | |
Recombinant Protein | |
E. coli | |
MLFRLSEHSS PEEEASPHQR ASGEGHHLKS KRPNPCAYTP PSLKAVQRIA ESHLQSISNL NENQASEEED ELGELRELGY PREEDEEEEE DDEEEEEEED SQAEVLKVIR QSAGQKTTCG QGLEGPWERP PPLDESERDG GSEDQVEDPA LSEPGEEPQR PSPSEPGT | |
Greater than 80% as determined by SDS-PAGE. |