missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein-lysine 6-oxidase Protein
A DNA sequence encoding the Homo sapiens (Human) Protein-lysine 6-oxidase, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_002308.2 |
---|---|
Gene ID (Entrez) | 4015 |
Name | Protein-lysine 6-oxidase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036961
|
enQuireBio™
QP8882-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16056961
|
enQuireBio™
QP8882-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16046961
|
enQuireBio™
QP8882-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16066961
|
enQuireBio™
QP8882-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16076961
|
enQuireBio™
QP8882-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16086961
|
enQuireBio™
QP8882-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_002308.2 | |
Protein-lysine 6-oxidase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4015 | |
Research Use Only | |
Lox | |
Recombinant Protein | |
E. coli | |
PYKYSDDNPY YNYYDTYERP RPGGRYRPGY GTGYFQYGLP DLVADPYYIQ ASTYVQKMSM YNLRCAAEEN CLASTAYRAD VRDYDHRVLL RFPQRVKNQG TSDFLPSRPR YSWEWHSCHQ HYHSMDEFSH YDLLDANTQR RVAEGHKASF CLEDTSCDYG YHRRFACTAH TQGLSPGCYD TYGADIDCQW IDITDVKPGN YILKVSVNPS YLVPESDYTN NVVRCDIRYT GHHAYASGCT ISPY | |
Greater than 90% as determined by SDS-PAGE. |