missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein-arginine deiminase type-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein-arginine deiminase type-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8239-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023491

  • 1643.42€ / 500µg

Estimated Shipment: 13-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_037490.2
Protein-arginine deiminase type-1 Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
29943
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PADI1
Recombinant Protein
Yeast
MAPKRVVQLS LKMPTHAVCV VGVEAHVDIH SDVPKGANSF RVSGSSGVEV FMVYNRTRVK EPIGKARWPL DTDADMVVSV GTASKELKDF KVRVSYFGEQ EDQALGRSVL YLTGVDISLE VDTGRTGKVK RSQGDKKTWR WGPEGYGAIL LVNCDRDNHR SAEPDLTHSW LMSLADLQDM SPMLLSCNGP DKLFDSHKLV LNVPFSDSKR VRVFCARGGN SLSDYKQVLG PQCLSYEVER QPGEQEIKFY VEGLTFPDAD FLGLVSLSVS LVDPGTLPEV TLFTDTVGFR MAPWIMTPNT QPPEELYVCR VMDTHGSNEK FLEDMSYLTL KANCKLTICP QVENRNDRWI QDEMEFGYIE APHKSFPVVF DSPRNRGLKD FPYKRILGPD FGYVTREIPL PGPSSLDSFG NLDVSPPVTV GGTEYPLGRI LIGSSFPKSG GRQMARAVRN FLKAQQVQAP VELYSDWLSV GHVDEFLTFV PTSDQKGFRL LLASPSACLK LFQEKKEEGY GEAAQFDGLK HQAKRSINEM LADRHLQRDN LHAQKCIDWN RNVLKRELGL AESDIVDIPQ LFFLKNFYAE AFFPDMVNMV VLGKYLGIPK PYGPIINGRC CLEEKVQSLL EPLGLHCIFI DDYLSYHELQ GEIHCGTNVR RKPFPFKWWN MVP
Videos
SDS
Documents
Certificates
Special Offers

Special Offers