missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein-arginine deiminase type-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein-arginine deiminase type-1, was expressed in the hosts and tags indicated.
202.00€ - 2155.00€
Specifications
Accession Number | NP_037490.2 |
---|---|
Gene ID (Entrez) | 29943 |
Name | Protein-arginine deiminase type-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083481
|
enQuireBio™
QP8239-YE-100UG |
100 μg |
645.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16023491
|
enQuireBio™
QP8239-YE-500UG |
500 μg |
1740.00€
500µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16093481
|
enQuireBio™
QP8239-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16013491
|
enQuireBio™
QP8239-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16033491
|
enQuireBio™
QP8239-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16003491
|
enQuireBio™
QP8239-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_037490.2 | |
Protein-arginine deiminase type-1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
29943 | |
Research Use Only | |
PADI1 | |
Recombinant Protein | |
Yeast | |
MAPKRVVQLS LKMPTHAVCV VGVEAHVDIH SDVPKGANSF RVSGSSGVEV FMVYNRTRVK EPIGKARWPL DTDADMVVSV GTASKELKDF KVRVSYFGEQ EDQALGRSVL YLTGVDISLE VDTGRTGKVK RSQGDKKTWR WGPEGYGAIL LVNCDRDNHR SAEPDLTHSW LMSLADLQDM SPMLLSCNGP DKLFDSHKLV LNVPFSDSKR VRVFCARGGN SLSDYKQVLG PQCLSYEVER QPGEQEIKFY VEGLTFPDAD FLGLVSLSVS LVDPGTLPEV TLFTDTVGFR MAPWIMTPNT QPPEELYVCR VMDTHGSNEK FLEDMSYLTL KANCKLTICP QVENRNDRWI QDEMEFGYIE APHKSFPVVF DSPRNRGLKD FPYKRILGPD FGYVTREIPL PGPSSLDSFG NLDVSPPVTV GGTEYPLGRI LIGSSFPKSG GRQMARAVRN FLKAQQVQAP VELYSDWLSV GHVDEFLTFV PTSDQKGFRL LLASPSACLK LFQEKKEEGY GEAAQFDGLK HQAKRSINEM LADRHLQRDN LHAQKCIDWN RNVLKRELGL AESDIVDIPQ LFFLKNFYAE AFFPDMVNMV VLGKYLGIPK PYGPIINGRC CLEEKVQSLL EPLGLHCIFI DDYLSYHELQ GEIHCGTNVR RKPFPFKWWN MVP | |
Greater than 90% as determined by SDS-PAGE. |