missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Programmed cell death protein 10 Protein

A DNA sequence encoding the Homo sapiens (Human) Programmed cell death protein 10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8043-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022341

Quantity Price
1 493.03€ / 100µg
6 463.44€ / 100µg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_009148.2
Programmed cell death protein 10 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
11235
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PDCD10
Recombinant Protein
E. coli
MRMTMEEMKN EAETTSMVSM PLYAVMYPVF NELERVNLSA AQTLRAAFIK AEKENPGLTQ DIIMKILEKK SVEVNFTESL LRMAADDVEE YMIERPEPEF QDLNEKARAL KQILSKIPDE INDRVRFLQT IKDIASAIKE LLDTVNNVFK KYQYQNRRAL EHQKKEFVKY SKSFSDTLKT YFKDGKAINV FVSANRLIHQ TNLILQTFKT VA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers