missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Poly(A)-specific ribonuclease PARN Protein
A DNA sequence encoding the Homo sapiens (Human) Poly(A)-specific ribonuclease PARN, was expressed in the hosts and tags indicated.
202.00€ - 2155.00€
Specifications
Accession Number | NP_001127949.1 |
---|---|
Gene ID (Entrez) | 5073 |
Name | Poly(A)-specific ribonuclease PARN Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023510
|
enQuireBio™
QP6467-YE-100UG |
100 μg |
645.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16063510
|
enQuireBio™
QP6467-YE-500UG |
500 μg |
1740.00€
500µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16033510
|
enQuireBio™
QP6467-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16053510
|
enQuireBio™
QP6467-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16073510
|
enQuireBio™
QP6467-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16043510
|
enQuireBio™
QP6467-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001127949.1 | |
Poly(A)-specific ribonuclease PARN Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5073 | |
Research Use Only | |
PARN | |
Recombinant Protein | |
Yeast | |
MEIIRSNFKS NLHKVYQAIE EADFFAIDGE FSGISDGPSV SALTNGFDTP EERYQKLKKH SMDFLLFQFG LCTFKYDYTD SKYITKSFNF YVFPKPFNRS SPDVKFVCQS SSIDFLASQG FDFNKVFRNG IPYLNQEEER QLREQYDEKR SQANGAGALS YVSPNTSKCP VTIPEDQKKF IDQVVEKIED LLQSEENKNL DLEPCTGFQR KLIYQTLSWK YPKGIHVETL ETEKKERYIV ISKVDEEERK RREQQKHAKE QEELNDAVGF SRVIHAIANS GKLVIGHNML LDVMHTVHQF YCPLPADLSE FKEMTTCVFP RLLDTKLMAS TQPFKDIINN TSLAELEKRL KETPFNPPKV ESAEGFPSYD TASEQLHEAG YDAYITGLCF ISMANYLGSF LSPPKIHVSA RSKLIEPFFN KLFLMRVMDI PYLNLEGPDL QPKRDHVLHV TFPKEWKTSD LYQLFSAFGN IQISWIDDTS AFVSLSQPEQ VKIAVNTSKY AESYRIQTYA EYMGRKQEEK QIKRKWTEDS WKEADSKRLN PQCIPYTLQN HYYRNNSFTA PSTVGKRNLS PSQEEAGLED GVSGEISDTE LEQTDSCAEP LSEGRKKAKK LKRMKKELSP AGSISKNSPA TLFEVPDTW | |
Greater than 90% as determined by SDS-PAGE. |